Skip to Content
Merck

Skip To

B4397

Bradykinin Fragment 1-8 acetate salt hydrate

≥97% (HPLC)

Synonym(s):

[des-Arg9]-Bradykinin acetate salt hydrate

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C44H61N11O10 · xC2H4O2 · yH2O
Molecular Weight:
904.02 (anhydrous free base basis)
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Assay:
≥97% (HPLC)
Form:
powder


assay

≥97% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

NC(CCCNC(N)=N)C(=O)N1CCCC1C(=O)N2CCCC2C(=O)NCC(=O)NC(Cc3ccccc3)C(=O)NC(CO)C(=O)N4CCCC4C(=O)NC(Cc5ccccc5)C(O)=O

InChI

1S/C44H61N11O10/c45-29(15-7-19-48-44(46)47)40(61)55-22-10-18-35(55)42(63)54-21-8-16-33(54)38(59)49-25-36(57)50-30(23-27-11-3-1-4-12-27)37(58)52-32(26-56)41(62)53-20-9-17-34(53)39(60)51-31(43(64)65)24-28-13-5-2-6-14-28/h1-6,11-14,29-35,56H,7-10,15-26,45H2,(H,49,59)(H,50,57)(H,51,60)(H,52,58)(H,64,65)(H4,46,47,48)

InChI key

VCEHWDBVPZFHAG-UHFFFAOYSA-N

Gene Information

General description

Bradykinin Fragment 1-8, also known as des-Arg9-bradykinin, is produced by the cleavage of bradykinin at 8-9 position by carboxypeptidase N.[1]

Application

Bradykinin Fragment 1-8 acetate salt hydrate has been used as a Bradykinin 1 receptor (B1R) agonist to investigate the expression and the role of B1R, in mediating neuronal injury under the chemical neurotoxicity paradigm in PC12 cell lines.[2] It has also been used as an analyte in nanostructure-initiator mass spectrometry and matrix-assisted laser desorption/ionization (MALDI).[3]

Biochem/physiol Actions

Bradykinin Fragment 1-8 is a selective B1 bradykinin receptor agonist.[4] It promotes the release of nitric oxide, prostacyclin, endothelium-derived hyperpolarizing factor in the endothelium. des-Arg9-bradykinin also promotes vasodilatation and increases blood flow in the peripheral circulation.[5]


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters