Skip to Content
Merck
All Photos(2)

Key Documents

B4032

Sigma-Aldrich

Banana powder

suitable for plant cell culture

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$137.00

$137.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$137.00

About This Item

UNSPSC Code:
10171502
NACRES:
NA.72

$137.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

form

powder

Quality Level

composition

Banana solids, 50 wt. %

technique(s)

cell culture | plant: suitable

application(s)

agriculture

storage temp.

2-8°C

Application

Banana extract B4032 is a mixture of banana puree and maltodextrin used to supplement plant growth media. It is used to promote root formation, plantlet strengthening and replating efficiency. Maltodextrose adds carbohydrates without the osmotic impact of mono- and disaccharides.

Other Notes

A spray dried mixture of banana puree and maltodextrin.

Reconstitution

Use at 30-80 g/L.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Chang et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 29(4), 313-317 (2005-02-15)
To select optimal media and conditions for the tissue culture of Dendrobium lituiforum. The basic media, light illumination condition, phytohormone, and natural aqueous extracts. were tested and compared. The chlorophyll contents and soluble protein contents were measured. N6 medium was
H Liu et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 23(11), 654-656 (2001-10-16)
To find a suitable medium for plantlet production of Dendrobium candidum in the period of strengthening plantlet. The basic medium, natural aqueous extracts, density of banana aqueous extract and of plant hormones were tested. The B5 or 1/2MS, added with
Songjun Zeng et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 27(9), 625-627 (2005-02-12)
The asepsis sowing and tissue culture of Bletilla striata were studied. The results indicated that the embyro culture had highest sprouting percent and plantlets percent when their embryos were mature. The optimal medium for the embryo culture was 1/2 MS.

Protocols

Preparation from Packaged Powder, Preparation from Basal Salt Solutions, Banana Powder Preparation and Use, Coconut Water Preparation and Use

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service