Skip to Content
Merck
All Photos(1)

Key Documents

B3656

Sigma-Aldrich

6-Benzylaminopurine-8-14C

aqueous ethanol solution

Synonym(s):

N6-Benzyladenine-8-14C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N5
CAS Number:
Molecular Weight:
225.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

aqueous ethanol solution

extent of labeling

40-60 mCi per mmol

shipped in

dry ice

storage temp.

−20°C

SMILES string

C(Nc1ncnc2nc[nH]c12)c3ccccc3

Packaging

sealed ampule

Physical form

Ethanol: water (9:1) solution

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicola Dikow et al.
American journal of medical genetics. Part A, 164A(12), 3061-3068 (2014-09-27)
Small interstitial deletions affecting chromosome region 3p25.3 have been reported in only five patients so far, four of them with overlapping telomeric microdeletions 3p25.3 and variable features of 3p- syndrome, and one patient with a small proximal microdeletion and a
Larisa Estefanía Cybulski et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(20), 6353-6358 (2015-05-06)
DesK is a bacterial thermosensor protein involved in maintaining membrane fluidity in response to changes in environmental temperature. Most likely, the protein is activated by changes in membrane thickness, but the molecular mechanism of sensing and signaling is still poorly
Yusuke Matsuzaka et al.
Molecular pharmacology, 88(6), 1085-1092 (2015-09-25)
Biliary excretion of organic anions, such as bile acids (BAs), is the main osmotic driving force for bile formation, and its impairment induces intrahepatic cholestasis. We investigated the involvement of Atp11c in the hepatic transport of organic anions using Atp11c
Bernd Wicklein et al.
Nature nanotechnology, 10(3), 277-283 (2014-11-05)
High-performance thermally insulating materials from renewable resources are needed to improve the energy efficiency of buildings. Traditional fossil-fuel-derived insulation materials such as expanded polystyrene and polyurethane have thermal conductivities that are too high for retrofitting or for building new, surface-efficient
Sepideh Amin-Hanjani et al.
Stroke, 46(7), 1850-1856 (2015-05-16)
Atherosclerotic vertebrobasilar disease is an important cause of posterior circulation stroke. To examine the role of hemodynamic compromise, a prospective multicenter study, Vertebrobasilar Flow Evaluation and Risk of Transient Ischemic Attack and Stroke (VERiTAS), was conducted. Here, we report clinical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service