Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 mg | Check Cart for Availability | $136.00 |
About This Item
biological source
synthetic (organic)
Quality Segment
assay
≥98% (TLC)
form
solid
color
dark red
solubility
DMSO: soluble, methanol: soluble
storage temp.
−20°C
SMILES string
O=C1NC(=O)C(c2c[nH]c3ccccc23)=C1c4c[nH]c5ccccc45
InChI
1S/C20H13N3O2/c24-19-17(13-9-21-15-7-3-1-5-11(13)15)18(20(25)23-19)14-10-22-16-8-4-2-6-12(14)16/h1-10,21-22H,(H,23,24,25)
InChI key
DQYBRTASHMYDJG-UHFFFAOYSA-N
Gene Information
human ... CDK2(1017), EGFR(1956)
rat ... Prkca(24680)
Biochem/physiol Actions
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥98% (TLC) | assay ≥98% (TLC) | assay ≥90% | assay 96% (TLC) |
| form solid | form solid | form solid | form solid |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| solubility DMSO: soluble, methanol: soluble | solubility DMSO: soluble | solubility DMSO: soluble | solubility DMSO: soluble |
| color dark red | color - | color - | color - |
Still not finding the right product?
Explore all of our products under Bisindolylmaleimide IV
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



