Skip to Content
Merck
All Photos(2)

Key Documents

B2188

Sigma-Aldrich

Anti-BPI (61-75) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Bactericidal permeability-increasing protein precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~54 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:1,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BPI(671)

General description

BPI (Bactericidal permeability-increasing protein) gene is mapped on to human chromosome 20q11.23. It belongs to the family of lipid-transfer proteins. BPI is mainly present in the granules of neutrophils and on the surface of neutrophils and monocytes.

Immunogen

synthetic peptide corresponding to amino acids 61-75 of human BPI

Application

Anti-BPI (61-75) antibody produced in rabbit has been used in immunoprecipitation.

Biochem/physiol Actions

BPI gene encodes a membrane associated bactericidal permeability increasing protein. BPI possesses high affinity for lipopolysaccharide. It has antimicrobial activity against gram-negative organisms and is essential constituent of the innate immune system for destroying the microbes as well as modulates subsequent adaptive immune responses. The amino-terminal of BPI is responsible for antimicrobial cytotoxicity and endotoxin-neutralization. The carboxyl-terminal is mainly for BPI-dependent transfer of Gram-negative bacteria and cell free endotoxin-rich particles to specific host cells. Mutation in BPI gene or a decrease of plasma BPI level may lead to chronic obstructive pulmonary disease (acute pneumonia and cystic fibrosis).

Target description

BPI (61-75), bactericidal/permeability-increasing protein, encodes a lipopolysaccharide binding protein. It is associated with human neutrophil granules and has bactericidal activity on gram-negative organisms.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karen McQuillan et al.
Frontiers in pharmacology, 11, 1098-1098 (2020-08-09)
In people with cystic fibrosis (PWCF), inflammation with concurrent infection occurs from a young age and significantly influences lung disease progression. Studies indicate that neutrophils are important effector cells in the pathogenesis of CF and in the development of anti-neutrophil
A myeloperoxidase-containing complex regulates neutrophil elastase release and actin dynamics during NETosis
Metzler KD, et al.
Testing, 8(3), 883-896 (2014)
Hendrik Schultz et al.
Clinica chimica acta; international journal of clinical chemistry, 384(1-2), 12-23 (2007-08-07)
Gram-negative bacteria (GNB) and their endotoxin present a constant environmental challenge. Endotoxins can potently signal mobilization of host defenses against invading GNB but also potentially induce severe pathophysiology, necessitating controlled initiation and resolution of endotoxin-induced inflammation to maintain host integrity.
Kathleen D Metzler et al.
Cell reports, 8(3), 883-896 (2014-07-30)
Neutrophils contain granules loaded with antimicrobial proteins and are regarded as impermeable organelles that deliver cargo via membrane fusion. However, during the formation of neutrophil extracellular traps (NETs), neutrophil elastase (NE) translocates from the granules to the nucleus via an
Alexander Holweg et al.
Biochemical Society transactions, 39(4), 1045-1050 (2011-07-27)
The human BPI (bactericidal/permeability-increasing protein), stored in primary azurophilic granula of neutrophil granulocytes and produced by mucosal epithelia, has been known for decades to bind LPS (lipopolysaccharide) with very high affinity and to efficiently kill Gram-negative bacteria. Thus BPI potentially

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service