Skip to Content
Merck

Skip To

B2017

Boc-Ala-Pro-Nva p-chlorothiobenzyl ester

≥98% (TLC), powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C25H36ClN3O5S
CAS Number:
Molecular Weight:
526.09
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Boc-Ala-Pro-Nva p-chlorothiobenzyl ester,

assay

≥98% (TLC)

Quality Segment

form

powder

solubility

chloroform: 10 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CCCC(NC(=O)C1CCCN1C(=O)C(C)NC(=O)OC(C)(C)C)C(=O)SCc2ccc(Cl)cc2

InChI

1S/C25H36ClN3O5S/c1-6-8-19(23(32)35-15-17-10-12-18(26)13-11-17)28-21(30)20-9-7-14-29(20)22(31)16(2)27-24(33)34-25(3,4)5/h10-13,16,19-20H,6-9,14-15H2,1-5H3,(H,27,33)(H,28,30)

InChI key

ILVSMEXNXGFDPM-UHFFFAOYSA-N

General description

Leucocyte elastase substrate

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M9771T2803M0883
assay

≥98% (TLC)

assay

≥98% (HPLC)

assay

≥98% (TLC)

assay

≥98% (TLC)

solubility

chloroform: 10 mg/mL, clear, colorless

solubility

chloroform: 50 mg/mL, clear, colorless

solubility

chloroform: 100 mg/mL, clear, colorless (Soluble in chloroform, methanol, and DMSO.)

solubility

chloroform: 100 mg/mL, clear, colorless

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

100

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters