Skip to Content
Merck
All Photos(2)

Key Documents

B1422

Sigma-Aldrich

B-Trisaccharide

Synonym(s):

α-D-Gal-(1→3)-(α-L-Fuc-[1→2])-D-Gal, Blood group B trisaccharide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32O15
CAS Number:
Molecular Weight:
488.44
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

Assay

≥95% (H-NMR)

Quality Level

form

solid

storage temp.

2-8°C

SMILES string

C[C@@H]1O[C@@H](O[C@H]2C(O)O[C@H](CO)[C@H](O)[C@@H]2O[C@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@@H]1O

InChI

1S/C18H32O15/c1-4-7(21)10(24)12(26)17(29-4)33-15-14(9(23)6(3-20)30-16(15)28)32-18-13(27)11(25)8(22)5(2-19)31-18/h4-28H,2-3H2,1H3/t4-,5+,6+,7+,8-,9-,10+,11-,12-,13+,14-,15+,16?,17-,18+/m0/s1

InChI key

XNBZPOHDTUWNMW-OUUCXATCSA-N

Application

Blood group B trisaccharide may be used to isolate, identify and characterize blood group trisaccharide specific lectins and ligands such as Langerin and MOA (Marasmius oreades agglutinin).[1][2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elin M Grahn et al.
Journal of molecular biology, 390(3), 457-466 (2009-05-12)
MOA (Marasmius oreades agglutinin), a lectin isolated from fruiting bodies of the mushroom M. oreades, specifically binds nonreducing terminal Galalpha(1,3)Gal carbohydrates, such as that which occurs in the xenotransplantation epitope Galalpha(1,3)Galbeta(1,4)GlcNAc and the branched blood group B determinant Galalpha(1,3)[Fucalpha(1,2)]Gal. Here
N O Seto et al.
European journal of biochemistry, 234(1), 323-328 (1995-11-15)
A 1034-bp synthetic gene encoding the human blood group B glycosyltransferase, which catalyzes the transfer of galactose from UDP-Gal to Fuc alpha(1-2)Gal beta-OR to give the blood group B determinant Gal alpha(1-3)[Fuc alpha(1-2)]Gal beta-OR (where R is a glycoprotein or
Ming Tan et al.
PloS one, 4(4), e5058-e5058 (2009-04-02)
Human noroviruses are the major viral pathogens of epidemic acute gastroenteritis. These genetically diverse viruses comprise two major genogroups (GI and GII) and approximately 30 genotypes. Noroviruses recognize human histo-blood group antigens (HBGAs) in a diverse, strain-specific manner. Recently the
Michiel van der Vlist et al.
Immunology and cell biology, 88(4), 410-415 (2010-03-24)
Langerhans cells (LC) are the first dendritic cells (DC) to encounter pathogens entering the body via mucosa or skin. Equipped with pattern recognition receptors (PRR), LC are able to detect and respond to these pathogens. An important PRR, in human
L Rydberg et al.
Sub-cellular biochemistry, 32, 107-125 (1999-07-07)
alpha-Gal terminated saccharides are present on the cell surface both as glycolipids and glycoproteins in all mammals except Old World monkeys and humans. The structural diversity among identified saccharides terminated by this epitope in animal tissues is steadily increasing. The

Articles

O-Linked glycoproteins are usually large proteins with a molecular mass of >200 kDa. Glycosylation generally occurs in high-density clusters and may represent as much as 50-80% of the overall mass.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service