B1127
Boron trifluoride-methanol solution
14% in methanol
Sign Into View Organizational & Contract Pricing
Select a Size
All Photos(1)
Select a Size
Change View
About This Item
Linear Formula:
BF3 · MeOH
CAS Number:
Molecular Weight:
99.85
Beilstein:
3611499
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Recommended Products
reaction suitability
reagent type: catalyst
core: boron
concentration
14% in methanol
storage temp.
2-8°C
SMILES string
CO.FB(F)F
InChI
1S/CH4O.BF3/c1-2;2-1(3)4/h2H,1H3;
InChI key
JBXYCUKPDAAYAS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
Sealed ampule.
Caution
Shelf life is extended considerably in sealed ampules.
Signal Word
Danger
Hazard Statements
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT RE 2 Inhalation - STOT SE 1
Target Organs
Kidney
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
60.8 °F - closed cup
Flash Point(C)
16 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
C Hue-Beauvais et al.
Journal of animal science, 93(4), 1641-1655 (2015-05-29)
Alterations to the metabolic endocrine environment during early life are crucial to mammary gland development. Among these environmental parameters, the initial nutritional event after birth is the consumption of milk, which represents the first maternal support provided to mammalian newborns.
Leila Nejadsadeghi et al.
Molecular biology reports, 42(2), 363-372 (2014-10-01)
Plant cells often increase cold tolerance by reprogramming their genes expression which results in adjusted metabolic alternations, a process enhanced under cold acclimation. In present study, we assessed the changes of membrane fatty acid compositions along with physio-biochemical indices like
Shulin Xiang et al.
Journal of pineal research, 59(1), 60-69 (2015-04-11)
Chemotherapeutic resistance, particularly to doxorubicin (Dox), represents a major impediment to successfully treating breast cancer and is linked to elevated tumor metabolism and tumor over-expression and/or activation of various families of receptor- and non-receptor-associated tyrosine kinases. Disruption of circadian time
Tao Wang et al.
Molecular medicine reports, 12(2), 3200-3206 (2015-05-13)
The aim of the present study was to investigate the effects of dietary trans fatty acids in mice. Following the administration of a 0.5/100 g diet of trans-9 octadecenoic acid (EA), trans-11 vaccenic acid (TVA) or cis-9, trans-11 conjugated linoleic
Nancy Hachicho et al.
PloS one, 10(8), e0134755-e0134755 (2015-08-21)
Morphology and physiology of fish embryos undergo dramatic changes during their development until the onset of feeding, supplied only by endogenous yolk reserves. For obtaining an insight how these restructuring processes are reflected by body mass related parameters, dry weights
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service