AXYTF50RS
Pipette filter tips
universal fit, racked, size 50 μL, pkg of 5x960tips/cs, sterile
Synonym(s):
pipet tips
About This Item
RNase free
barrier: no
endotoxin free
filter
Corning
Recommended Products
material
clear
sterility
sterile
feature
DNA free
RNase free
barrier: no
endotoxin free
filter
manufacturer/tradename
Axygen Inc TF-50-R-S
Corning
packaging
pkg of 5x960tips/cs
size
50 μL
volume
50 μL
Looking for similar products? Visit Product Comparison Guide
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service