AXYSCT200SSV
Screw cap tubes and caps with O-rings, self-standing
size 2.0 mL, violet, pkg of 8x500caps/cs
Synonym(s):
centrifuge tubes
About This Item
manufacturer/tradename
Axygen Inc SCT-200-SS-V
Corning
packaging
pkg of 8x500caps/cs
size
2.0 mL
color
violet
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service