Key Documents
AXYPDW500C
Deep well plate
for SBS FootPrint, size 96 wells, size 600 μL, pkg of 10x5plates/cs
Synonym(s):
Corning cell culture plate
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material:
conical bottom wells
size:
600 μL
96 wells
Pricing and availability is not currently available.
material
conical bottom wells
manufacturer/tradename
Axygen Inc P-DW-500-C
Corning
packaging
pkg of 10x5plates/cs
size
600 μL
96 wells
General description
  • Axygen® deep well assay plates are ideal for use in applications including sample collection, in vitro growth chambers and long-term storage.
  • Chemically resistant and withstands temperatures down to -80°C.
  • Ultra-low profile design for reduced space requirements.
  • Flat surface for efficient sealing using heat sealing film.
Legal Information
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)