Skip to Content
Merck
All Photos(1)

Key Documents

AV50878

Sigma-Aldrich

Anti-MDFIC (AB2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-HIC, Anti-MyoD family inhibitor domain containing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

37 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MDFIC(29969)

Immunogen

Synthetic peptide directed towards the middle region of human MDFIC

Application

Anti-MDFIC (AB2) antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.

Biochem/physiol Actions

MyoD family inhibitor domain containing (MDFIC) is involved in transcription regulation of viral genome expression. Alternate translation results in two isoforms, p40 and p32 found localized to cytoplasm and nucleolus, respectively.[1]

Sequence

Synthetic peptide located within the following region: PQLQTSAQVPSGEEIGKIKNGHTGLSNGNGIHHGAKHGSADNRKLSAPVS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ella Reiss-Sklan et al.
PloS one, 4(7), e6152-e6152 (2009-07-08)
Human I-mfa domain containing protein (HIC) differentially regulates transcription from viral promoters. HIC affects the Wnt pathway, the JNK/SAPK pathway and the activity of positive transcription elongation factor-b (P-TEFb). Studies exploring HIC function in mammalian cells used ectopically expressed HIC
Lili Gu et al.
Immunology letters, 123(2), 179-184 (2009-05-12)
The Human I-mfa domain-Containing protein, HIC, is a 246 amino acid protein that functions as a transcriptional regulator. Although the precise function of HIC remains to be clarified, the association of the HIC gene locus with myeloid neoplasms, its interactions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service