Skip to Content
Merck
All Photos(1)

Key Documents

AV50642

Sigma-Aldrich

Anti-NRIP3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-C11orf14, Anti-NY-SAR-105, Anti-Nuclear receptor interacting protein 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

27 kDa

species reactivity

guinea pig, rabbit, mouse, rat, dog, human, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NRIP3(56675)

General description

NRIP3 codes for nuclear receptor interacting protein 3. It may be involved in pediatric acute myeloid leukemia[1].
Rabbit Anti-NRIP3 antibody recognizes canine, bovine, human, mouse, and rat NRIP3.

Immunogen

Synthetic peptide directed towards the middle region of human NRIP3

Application

Rabbit Anti-NRIP3 antibody is suitable for western blot applications at a concentration of 0.5 μg/ml.

Biochem/physiol Actions

The exact functions of NRIP3 remain unknown.

Sequence

Synthetic peptide located within the following region: ALVDTGCLYNLISLACVDRLGLKEHVKSHKHEGEKLSLPRHLKVVGQIEH

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NRIP3: a novel translocation partner of MLL detected in a pediatric acute myeloid leukemia with complex chromosome 11 rearrangements.
Brian V Balgobind et al.
Haematologica, 94(7), 1033-1033 (2009-05-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service