Skip to Content
Merck
All Photos(3)

Key Documents

AV46802

Sigma-Aldrich

Anti-SILV antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-D12S53E, Anti-Gp100, Anti-ME20, Anti-PMEL17, Anti-SI, Anti-SIL, Anti-Silver homolog (mouse)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

70 kDa

species reactivity

mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SILV(6490)

Immunogen

Synthetic peptide directed towards the N terminal region of human SILV

Application

Anti-SILV antibody produced in rabbit is suitable for western blotting at a concentration of 1.25μg/mL. It is also useful for immunohistochemistry at a concentration of 4-8μg/mL.

Biochem/physiol Actions

SILV [Silver homolog (mouse)] gene encodes a melanocyte-specific type I transmembrane glycoprotein expressed primarily in melanocytes and at low levels in normal cell lines and tissues. It facilitates in the structural organization of premelanosomes within multivesicular bodies. The encoded protein is also involved in generating internal matrix fibers that define the transition from stage I to stage II melanosomes.

Sequence

Synthetic peptide located within the following region: HFLRNQPLTFALQLHDPSGYLAEADLSYTWDFGDSSGTLISRALVVTHTY

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Bailin et al.
The Journal of investigative dermatology, 106(1), 24-27 (1996-01-01)
We have determined the DNA sequence and genomic organization of D12S53E (Pmel 17), the human homologue of the mouse silver (si) locus. D12S53E encodes a melanosomal matrix protein whose expression is closely correlated with cellular melanin content and which is
J F Berson et al.
Molecular biology of the cell, 12(11), 3451-3464 (2001-11-06)
Melanosomes are tissue-specific organelles within which melanin is synthesized and stored. The melanocyte-specific glycoprotein Pmel17 is enriched in the lumen of premelanosomes, where it associates with characteristic striations of unknown composition upon which melanin is deposited. However, Pmel17 is synthesized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service