Skip to Content
Merck
All Photos(1)

Key Documents

AV44138

Sigma-Aldrich

Anti-SLCO6A1 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-GST, Anti-MGC26949, Anti-OATP6A1, Anti-OATPY, Anti-Solute carrier organic anion transporter family, member 6A1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

79 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

Related Categories

Immunogen

Synthetic peptide directed towards the N terminal region of human SLCO6A1

Application

Anti-SLCO6A1 (AB1) antibody produced in rabbit is suitable for western blotting at a concentration of 5.0μg/ml.

Biochem/physiol Actions

SLCO6A1 (GST) is an organic anion transporter expressed strongly in testis and weakly in brain (adult and fetal), placenta and spleen. It is a putative cancer/testis (CT) cell surface antigen that might serve as a marker in medulloblastoma.

Sequence

Synthetic peptide located within the following region: CCNNIRCFMIFYCILLICQGVVFGLIDVSIGDFQKEYQLKTIEKLALEKS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sueli M Oba-Shinjo et al.
Cancer immunity, 8, 7-7 (2008-04-23)
Medulloblastoma is the most common childhood malignant tumor of the central nervous system. Treatment of medulloblastoma requires harmful therapy and nevertheless carries a poor prognosis. Due to their presence in various cancers and their limited expression in normal tissues, CT
Sang-Yull Lee et al.
Cancer immunity, 4, 13-13 (2004-11-18)
Serological analysis of recombinant cDNA expression libraries (SEREX) has led to the identification of many of the antigens recognized by the immune system of cancer patients, which are collectively referred to as the cancer immunome. We used SEREX to screen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service