Skip to Content
Merck
All Photos(1)

Key Documents

AV40654

Sigma-Aldrich

Anti-SARS (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Seryl-tRNA synthetase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

57 kDa

species reactivity

bovine, horse, mouse, guinea pig, rat, rabbit, human, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SARS(6301)

General description

tRNAs are aminoacylated by the aminoacyl-tRNA synthetases. Due to redundancy of the genetic code, which allows for 64 mRNA codons, tRNA isoacceptors exist that may be aminoacylated with one amino acid but differ in their anticodons. Seryl-tRNA synthetase (SARS) is an enzyme that aminoacylates target tRNA with serine. Seryl-tRNA-synthetase interacts with the tRNA(Ser) acceptor stem, which makes this part of the tRNA a valuable structural element for investigating motifs of the protein-RNA complex. Cytosolic seryl-tRNA synthetase (hsSerRS) is responsible for the covalent attachment of serine to its cognate tRNA(Ser).

The previously assigned protein identifier Q5T5C8 has been merged into P49591. Full details can be found on the UniProt database.

Specificity

Anti-SARS (AB1) polyclonal antibody reacts with canine, zebrafish, chicken, human, mouse, rat, and bovine cytosolic seryl-tRNA synthetases.

Immunogen

Synthetic peptide directed towards the C terminal region of human SARS

Application

Anti-SARS (AB1) polyclonal antibody is used to tag cytosolic seryl-tRNA synthetase for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of cytosolic seryl-tRNA synthetase in tRNA serine acylation and translation

Biochem/physiol Actions

SARS belongs to the class II amino-acyl tRNA family. The enzyme catalyzes the transfer of L-serine to tRNA (Ser) and is related to bacterial and yeast counterparts.This gene belongs to the class II amino-acyl tRNA family. The encoded enzyme catalyzes the transfer of L-serine to tRNA (Ser) and is related to bacterial and yeast counterparts.

Sequence

Synthetic peptide located within the following region: PEKLKEFMPPGLQELIPFVKPAPIEQEPSKKQKKQHEGSKKKAAARDVTL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service