Skip to Content
Merck
All Photos(1)

Key Documents

AV39289

Sigma-Aldrich

Anti-NKRF antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-NF-kappaB repressing factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

44 kDa

species reactivity

human, pig, horse, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NKRF(55922)

General description

NKRF codes for a nuclear protein that functions as a repressing factor for NF-κB. NKRF suppresses the stimulation of hiNOS, IL-8, IFN, and HIV-1 by NFκB.
Rabbit Anti-NKRF antibody recognizes bovine, human, mouse, rat, and canine NKRF.

Immunogen

Synthetic peptide directed towards the N terminal region of human NKRF

Application

Rabbit Anti-NKRF antibody is suitable for western blot applications at a concentration of 2.5 μg/ml.

Biochem/physiol Actions

NKRF is a transcription factor that interacts with specific negative regulatory elements (NREs) to mediate transcriptional repression of certain NK-kappa-B-responsive genes. NKRF localizes predominantly to the nucleolus with a small fraction found in the nucleoplasm and cytoplasm.This gene encodes a transcription factor that interacts with specific negative regulatory elements (NREs) to mediate transcriptional repression of certain NK-kappa-B-responsive genes. The protein localizes predominantly to the nucleolus with a small fraction found in the nucleoplasm and cytoplasm.

Sequence

Synthetic peptide located within the following region: MEKILQMAEGIDIGEMPSYDLVLSKPSKGQKRHLSTCDGQNPPKKQAGSK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gene expression profile of NFκB repressing factor (NKRF) knockdown cells by microarray analysis
Sun, Y., et al.
BioChip Journal, 6(3), 247-253 (2012)
Yung-Yang Liu et al.
PloS one, 8(6), e66760-e66760 (2013-07-11)
High-tidal-volume mechanical ventilation used in patients with acute lung injury (ALI) can induce the release of inflammatory cytokines, as macrophage inflammatory protein-2 (MIP-2), recruitment of neutrophils, and disruption of alveolar epithelial and endothelial barriers. Induced pluripotent stem cells (iPSCs) have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service