Skip to Content
Merck
All Photos(1)

Key Documents

AV38985

Sigma-Aldrich

Anti-MyST4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MyST Histone acetyltransferase (monocytic leukemia) 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

231 kDa

species reactivity

human, guinea pig, mouse, rabbit, dog, bovine, horse, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYST4(23522)

Immunogen

Synthetic peptide directed towards the N terminal region of human MYST4

Biochem/physiol Actions

MyST4 or KAT6B is a K(lysine) acetyltransferase that also exhibits transcriptional activation and repression activity. It is a component of MOZ/MORF protein complex. Anomalies in KAT6B gene lead to aberrations in developmental programs resulting in Genitopatellar syndrome (GPS) and Say-Barber-Biesecker-Young-Simpson syndrome (SBBYSS or Ohdo syndrome).

Sequence

Synthetic peptide located within the following region: MVKLANPLYTEWILEAIQKIKKQKQRPSEERICHAVSTSHGLDKKTVSEQ

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahrin Huh et al.
American journal of orthodontics and dentofacial orthopedics : official publication of the American Association of Orthodontists, its constituent societies, and the American Board of Orthodontics, 144(4), 568-576 (2013-10-01)
Genetic influences on the development of malocclusion include heritable effects on both masticatory muscles and jaw skeletal morphology. Beyond genetic variations, however, the characteristics of muscle and bone are also influenced by epigenetic mechanisms that produce differences in gene expression.
Philippe M Campeau et al.
American journal of human genetics, 90(2), 282-289 (2012-01-24)
Genitopatellar syndrome (GPS) is a skeletal dysplasia with cerebral and genital anomalies for which the molecular basis has not yet been determined. By exome sequencing, we found de novo heterozygous truncating mutations in KAT6B (lysine acetyltransferase 6B, formerly known as
Philippe M Campeau et al.
Human mutation, 33(11), 1520-1525 (2012-06-21)
Genitopatellar syndrome (GPS) and Say-Barber-Biesecker-Young-Simpson syndrome (SBBYSS or Ohdo syndrome) have both recently been shown to be caused by distinct mutations in the histone acetyltransferase KAT6B (a.k.a. MYST4/MORF). All variants are de novo dominant mutations that lead to protein truncation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service