Skip to Content
Merck
All Photos(2)

Documents

AV38881

Sigma-Aldrich

Anti-RXRG (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-NR2B3, Anti-RXRC, Anti-Retinoid X receptor, γ

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

51 kDa

species reactivity

guinea pig, rat, dog, human, mouse, rabbit, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RXRG(6258)

Immunogen

Synthetic peptide directed towards the C terminal region of human RXRG

Biochem/physiol Actions

RXRG belongs to the retinoid receptor family of nuclear receptors that mediate the cellular effects of retinoic acid. RXRG increases DNA binding and transcription of genes regulated by retinoic acid, thyroid hormone and vitamin D. Mutations in RXRG gene is reportedly associated with diabetic retinopathy.

Sequence

Synthetic peptide located within the following region: LEAYTKQKYPEQPGRFAKLLLRLPALRSIGLKCLEHLFFFKLIGDTPIDT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcia I Dawson et al.
Biochimica et biophysica acta, 1821(1), 21-56 (2011-10-25)
This chapter presents an overview of the current status of studies on the structural and molecular biology of the retinoid X receptor subtypes α, β, and γ (RXRs, NR2B1-3), their nuclear and cytoplasmic functions, post-transcriptional processing, and recently reported ligands.
C-H Hsieh et al.
Genetics and molecular research : GMR, 10(4), 3545-3551 (2011-12-20)
Retinoid-X receptor (RXR) is one of the members of the nuclear hormone receptor superfamily. It forms heterodimers with many nuclear receptors, such as the peroxisome proliferative-activated receptor, which has been proposed to be involved in diabetic complications, including retinopathy. A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service