Skip to Content
Merck
All Photos(2)

Key Documents

AV35151

Sigma-Aldrich

Anti-KCNAB3 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Potassium voltage-gated channel, shaker-related subfamily, β member 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

44 kDa

species reactivity

bovine, guinea pig, horse, rat, dog, rabbit, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... KCNAB3(9196)

General description

KCNAB3 codes for a member of potassium voltage-gated channel (shaker-related) protein subfamily. This protein is a β subunit and can form heterodimers that modulate the function of α subunits.
Rabbit Anti-KCNAB3 antibody recognizes rat, canine, human, bovine, and mouse KCNAB3.

Immunogen

Synthetic peptide directed towards the N terminal region of human KCNAB3

Application

Rabbit Anti-KCNAB3 antibody is suitable for western blot (0.65 μg/ml) and IHC (4-8 μg/ml) applications.

Biochem/physiol Actions

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes - shaker, shaw, shab, and shal - have been identified in Drosophila, and each has been shown to have human homolog(s). This gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily. This member is one of the beta subunits, which are auxiliary proteins associating with functional Kv-alpha subunits. This member and the KCNA5 gene product assemble into a heteromultimeric A-type channel that inactivates completely and is significantly faster than other A-type Kv channels.

Sequence

Synthetic peptide located within the following region: RNLGKSGLRVSCLGLGTWVTFGSQISDETAEDVLTVAYEHGVNLFDTAEV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service