Skip to Content
Merck
All Photos(3)

Key Documents

AV32765

Sigma-Aldrich

Anti-CDX4 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Caudal type homeobox transcription factor 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

30 kDa

species reactivity

horse, dog, rabbit, pig, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDX4(1046)

General description

Caudal related homeobox (Cdx) genes are known to modulate axial elongation and patterning in many vertebrate embryos. Cdx4 regulates the ontogenesis of placental labyrinth in mice. Furthermore, Cdx4 is known to dysregulate Hox gene expression which causes acute myeloid leukemia in mouse models.
Rabbit Anti-CDX4 antibody recognizes rat, mouse, bovine, and human CDX4.

Immunogen

Synthetic peptide directed towards the C terminal region of human CDX4

Application

Rabbit Anti-CDX4 (AB1) antibody can be used for western blot applications at a concentration of 2.5 μg/ml.

Biochem/physiol Actions

CDX are homeodomain transcription factors related to the Drosophila caudal gene. The vertebrate CDX have been implicated in the development of the posterior embryo. Several signaling molecules, notably retinoic acid (RA) and members of the Wnt (wingless) and fibroblast growth factor (FGF) families, are also implicated in patterning of the posterior vertebrate embryo. CDX family is the target of Wnt, RA and FGF signaling, suggesting that CDX factors act to convey the activity of these signaling molecules to Hox genes.

Sequence

Synthetic peptide located within the following region: KKISQFENSGGSVQSDSDSISPGELPNTFFTTPSAVRGFQPIEIQQVIVS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johan van Nes et al.
Development (Cambridge, England), 133(3), 419-428 (2006-01-07)
Caudal related homeobox (Cdx) genes have so far been shown to be important for embryonic axial elongation and patterning in several vertebrate species. We have generated a targeted mutation of mouse Cdx4, the third member of this family of transcription
Dimple Bansal et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(45), 16924-16929 (2006-10-28)
HOX genes have emerged as critical effectors of leukemogenesis, but the mechanisms that regulate their expression in leukemia are not well understood. Recent data suggest that the caudal homeobox transcription factors CDX1, CDX2, and CDX4, developmental regulators of HOX gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service