Skip to Content
Merck
All Photos(2)

Key Documents

AV31500

Sigma-Aldrich

Anti-TCFL5 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Transcription factor-like 5 (basic helix-loop-helix)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

53 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TCFL5(10732)

General description

TCFL5 is a basic helix-loop-helix (bHLH) protein that is expressed in primary spermatocytes. Studies in mice have revealed that Tcfl5 associated with the Calmegin gene promoter during spermatogenesis.
Rabbit Anti-TCFL5 antibody recognizes mouse and human TCFL5.

Immunogen

Synthetic peptide directed towards the middle region of human TCFL5

Application

Rabbit Anti-TCFL5 antibody can be used for western blot assays at a concentration of 2.0μg/ml. The antibody product can also be used for IHC applications (4-8μg/ml, using paraffin-embedded tissues).

Biochem/physiol Actions

TCFL5 is a new bHLH transcription factor that negatively regulates upstream transcription factor-dependent transcription.

Sequence

Synthetic peptide located within the following region: TLIRHPSELMNVPLQQQNKCTALVKNKTAATTTALQFTYPLFTTNACSTS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Maruyama et al.
Cytogenetics and cell genetics, 82(1-2), 41-45 (1998-10-09)
We have isolated a novel human gene that is expressed specifically in primary spermatocytes in the testis. The cDNA contains an open reading frame of 1356 bp, encoding a 452-amino-acid protein that includes a basic Helix-Loop-Helix (bHLH) motif. The gene
Michel Siep et al.
Nucleic acids research, 32(21), 6425-6436 (2004-12-09)
In mouse spermatogenesis, differentiating germ line cells initiate expression of specific genes at subsequent developmental steps. The Calmegin (Clgn) gene is first expressed in meiotic prophase, in primary spermatocytes, and encodes a protein that acts as a chaperone. To identify

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service