Skip to Content

AV30002

Anti-CBX4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Chromobox homolog 4 (Pc class homolog, Drosophila)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

61 kDa

species reactivity

rat, guinea pig, bovine, rabbit, dog, human, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CBX4(8535)

Immunogen

Synthetic peptide directed towards the N terminal region of human CBX4

Application

Anti-CBX4 antibody produced in rabbit is suitable for western blotting at a concentration of 0.5 μg/ml. For immunohistochemistry of paraffin-embedded tissue sections, a concentration of 4-8 μg/ml is suitable.

Biochem/physiol Actions

CBX4 belongs to the Polycomb group of proteins that are involved in determining stem cell pluripotency, identity and epigenetic gene silencing. CBX4 exhibits E3 sumo ligase activity that is directly involved in DNA damage response pathway. It interacts with p63 and regulates the transcription of genes that maintain the stemness of epithelial progenitors. CBX4 has a non-redundant role in T-cell proliferation and thymic organogenesis.

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Other Notes

Synthetic peptide located within the following region: LLIAFQNRERQEQLMGYRKRGPKPKPLVVQVPTFARRSNVLTGLQDSSTD

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library