Skip to Content
Merck
All Photos(1)

Documents

AV03047

Sigma-Aldrich

Anti-CDKN2B antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

15 kDa

species reactivity

human, pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDKN2B(1030)

Immunogen

Synthetic peptide directed towards the middle region of human CDKN2B

Application

Anti-CDKN2B antibody produced in rabbit is suitable for western blotting at a concentration of 1 μg/ml.

Biochem/physiol Actions

CDKN2B has inhibitory effects on cell cycle progression. It binds to cyclin-dependent kinases and prevents their association with D-type cyclins. Methylation of CDKN2B has been associated with pathogenesis of pediatric myelodysplastic syndromes and in malignant hematopoiesis. Mutations in CDKN2B gene have been observed in malignant gliomas.

Sequence

Synthetic peptide located within the following region: REGFLDTLVVLHRAGARLDVRDAWGRLPVDLAEERGHRDVAGYLRTATGD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miyoung Kim et al.
Acta haematologica, 130(2), 115-121 (2013-04-11)
Transcriptional repression of tumor suppressor genes is determined by the quantity of promoter hypermethylation. We analyzed the methylation quantity of CDKN2B in pediatric myelodysplastic syndromes (MDS). Quantitative measurement of CDKN2B methylation was performed in 25 pediatric MDS patients and 12
Utz Krug et al.
Oncogene, 21(21), 3475-3495 (2002-05-29)
Over the last decade, a growing number of tumor suppressor genes have been discovered to play a role in tumorigenesis. Mutations of p53 have been found in hematological malignant diseases, but the frequency of these alterations is much lower than
Yanhong Liu et al.
Current opinion in genetics & development, 20(3), 239-244 (2010-03-10)
Recent advances in human genome studies have opened new avenues for the identification of susceptibility genes for many complex genetic disorders, especially in the field of rare cancers such as glioma. To date, eight glioma susceptibility loci have been identified
Moon-Taek Park et al.
Journal of biochemistry and molecular biology, 36(1), 60-65 (2003-01-25)
Cancer is frequently considered to be a disease of the cell cycle. As such, it is not surprising that the deregulation of the cell cycle is one of the most frequent alterations during tumor development. Cell cycle progression is a
Nerea Méndez-Barbero et al.
EBioMedicine, 46, 274-289 (2019-08-10)
Tumor necrosis factor-like weak inducer of apoptosis (Tnfsf12; TWEAK) and its receptor Fibroblast growth factor-inducible 14 (Tnfrsf12a; Fn14) participate in the inflammatory response associated with vascular remodeling. However, the functional effect of TWEAK on vascular smooth muscle cells (VSMCs) is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service