Skip to Content
Merck
All Photos(2)

Key Documents

AV03041

Sigma-Aldrich

Anti-CDC25B (AB2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Cell division cycle 25 homolog B (S. pombe)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

65 kDa

species reactivity

human, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDC25B(994)

Immunogen

Synthetic peptide directed towards the N terminal region of human CDC25B

Application

Anti- CDC25B (AB2) antibody produced in rabbit is suitable for western blotting at a concentration of 1 μg/ml. For immunohistochemistry of paraffin-embedded tissue sections, a concentration of 4-8 μg/ml is suitable.

Biochem/physiol Actions

CDC25 phosphatases regulate the progression of cell cycle. The activity of CDC25B is important for G2/M transition, cell migration, invasion and survival. The PKA/CDC25B pathway plays a crucial role in meiotic arrest of mouse oocytes. The activity of CDC25B is regulated by 14-3-3 proteins.

Sequence

Synthetic peptide located within the following region: MEVPQPEPAPGSALSPAGVCGGAQRPGHLPGLLLGSHGLLGSPVRAAASS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takumi Kawabe
Molecular cancer therapeutics, 3(4), 513-519 (2004-04-14)
Many conventional anticancer treatments kill cells irrespective of whether they are normal or cancerous, so patients suffer from adverse side effects due to the loss of healthy cells. Anticancer insights derived from cell cycle research has given birth to the
Jun Meng et al.
PloS one, 8(1), e53633-e53633 (2013-01-18)
The protein kinase A (PKA)/Cdc25B pathway plays a critical role in maintaining meiotic arrest in mouse oocytes. However, the molecular mechanism underlying this interchange is not known. In this study, we assessed the role of 14-3-3ε interaction with phosphorylated Cdc25B
Xiu-Yue Yu et al.
Asian Pacific journal of cancer prevention : APJCP, 13(3), 931-935 (2012-05-29)
Cdc25 phosphatases are important regulators of the cell cycle. Their abnormal expression detected in a number of tumors implies that their dysregulation is involved in malignant transformation. However, the role of Cdc25B in renal cell carcinomas remains unknown. To shed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service