Skip to Content
Merck
All Photos(1)

Documents

AQ0898

AQUA Peptide YTLNILEDIGGGQ[K]

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116026

mol wt

monoisotopic mol wt 1527.80 Da

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Lcp1(18826)
rat ... Lcp1(306071)

Application

Protein-AQUA is a powerful, enabling technology that facilitates focused, quantitative studies of not only specific protein expression, but specific amino acid modification as well, using stable-isotope–labeled peptides and mass spectrometry.

Legal Information

This method was developed by Dr. Steve Gygi and colleagues at Harvard Medical School. Limited use of this method is permitted under a licensing arrangement with Harvard Medical School.
AQUA is a trademark of Harvard Medical School

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service