Skip to Content
Merck

Skip To

APPA002

Green Universal Detection Reagent

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 mL
Check Cart for Availability
$637.00

About This Item

NACRES:
NA.25
UNSPSC Code:
12352204

$637.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

technique(s)

microbe id | specific enzyme detection: suitable

shipped in

dry ice

storage temp.

−20°C

General description

GreenUniversal Detection Reagent is formulated with green fluorescent protein (GFP),which is a part of the split fluorescent protein (FP) system that may be usedto quantify the expression level of a tagged protein of interest (POI).

Application

The S1-10 Universal Detection Reagent is provided as a ready-to-use solution for use with the In Vitro "Fold ′n′ Glow" Solubility Assay Kits:

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RU5500GERPN3004DUO92014
technique(s)

microbe id | specific enzyme detection: suitable

technique(s)

immunofluorescence: suitable

technique(s)

-

technique(s)

proximity ligation assay: suitable

Quality Level

200

Quality Level

300

Quality Level

-

Quality Level

200

storage temp.

−20°C

storage temp.

-

storage temp.

2-8°C

storage temp.

−20°C

shipped in

dry ice

shipped in

2-8°C

shipped in

-

shipped in

dry ice

form

solution

form

-

form

-

form

-


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters