Skip to Content
Merck
All Photos(6)

Documents

AMAB91170

Sigma-Aldrich

Monoclonal Anti-LAMP1 antibody produced in mouse

Prestige Antibodies® Powered by Atlas Antibodies, clone CL3484, purified immunoglobulin, buffered aqueous glycerol solution

Synonym(s):

Anti-CD107a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CL3484, monoclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 1 μg/mL
immunohistochemistry: 1:5000- 1:10000

isotype

IgG2a

immunogen sequence

NFSAAFSVNYDTKSGPKNMTFDLPSDATVVLNRSSCGKENTSDPSLVIAFGRGHTLTLNFTRNATRYSVQLMSFVYNLSDTHLFPNASSKEIKTVESITDIRADIDKKYRCVSGTQVHMNNVTVTLHDA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LAMP1(3916)

General description

The gene LAMP1 (lysosomal-associated membrane protein 1) encodes a type I transmembrane protein has a short cytoplasmic tail containing a lysosome-targeting signal of GYQTI(382)-COOH. The gene is mapped to human chromosome 13q34.

Immunogen

lysosomal-associated membrane protein 1

Application

All Prestige Antibodies® Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene LAMP1 (lysosomal associated membrane protein 1) encodes a membrane glycoprotein that functions as an intracellular receptor. It is found to be expressed in the cytoplasm of several types of tumor cells and may be involved in tumor invasion. Lamp1 is crucial for perforin trafficking to the lytic granules and motility of these lytic granules. Its knockdown leads to inhibition of cytotoxicity of human natural killer cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST93498

Physical form

Phospate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acidic pH-Induced Conformations and LAMP1 Binding of the Lassa Virus Glycoprotein Spike.
Li S
PLoS Pathogens, 12, e1005418-e1005418 (2016)
Expression of the lysosomal-associated membrane protein-1 (LAMP-1) in astrocytomas.
Jensen SS
International Journal of Clinical and Experimental Pathology, 6, 1294-1305 (2013)
LAMP1/CD107a is required for efficient perforin delivery to lytic granules and NK-cell cytotoxicity.
Krzewski K
Blood, 121, 4672-4683 (2013)
Monosomy and ring chromosome 13 in a thyroid nodular goiter-do we underestimate its relevance in benign thyroid lesions?
Sendt W
Cancer Genetics, 205, 128-130 (2012)
Kenji Akasaki et al.
Journal of biochemistry, 156(1), 39-49 (2014-04-04)
Lysosome-associated membrane protein-1 (LAMP-1) is a type I transmembrane protein with a short cytoplasmic tail that possesses a lysosome-targeting signal of GYQTI(382)-COOH. Wild-type (WT)-LAMP-1 was exclusively localized in high density lysosomes, and efficiency of LAMP-1's transport to lysosomes depends on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service