Skip to Content
Merck
All Photos(7)

Key Documents

AMAB90911

Sigma-Aldrich

Monoclonal Anti-ITGAM antibody produced in mouse

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, clone CL1719, purified immunoglobulin, buffered aqueous glycerol solution

Synonym(s):

Anti-CD11B, Anti-CR3A, Anti-MAC-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CL1719, monoclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 1 μg/mL
immunohistochemistry: 1:1000- 1:2500

isotype

IgG1

immunogen sequence

LNFTASENTSRVMQHQYQVSNLGQRSLPISLVFLVPVRLNQTVIWDRPQVTFSENLSSTCHTKERLPSHSDFLAELRKAPVVNCSIAVCQRIQCDIPFFGIQEEFNATLKGNLSFDWYIKTSHNHLLIVSTAEIL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ITGAM(3684)

General description

Integrin α M (ITGAM) is also called as cluster of differentiation 11b (CD11b), intercellular adhesion molecule-1 (ICAM-1), complement component 3 receptor α chain (CR3α), macrophage-1 antigen α subunit or macrophage receptor 1 α subunit (MAC1α). It is expressed on monocytes/macrophages, natural killer cells, neutrophils and a subset of B-and T-lymphocytes. ITGAM gene is located on human chromosome 16p11.

Immunogen

integrin, alpha M (complement component 3 receptor 3 subunit)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Monoclonal Anti-ITGAM Prestige Antibodies® Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using protein array and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/ Prestige.

Biochem/physiol Actions

Integrin α M (ITGAM) is involved in cell activation, chemotaxis, cytotoxicity and phagocytosis. It modulates the interaction of leukemic cells with microenvironment by binding inactivated complement component 3b (iC3b), intercellular adhesion molecule (ICAM), fibrinogen, β-glukanes, coagulation factor X etc. ITGAM is considered as a marker for myeloid-derived suppressor cells that participates in restraining antitumor immunity of the host and inducing the enlargement and drug-resistance of hematological malignant cells. D11b integrin contributes to ICAM-1-dependent AIDP patient mononuclear leukocyte adhesion.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83070

Physical form

Phospate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A nonsynonymous functional variant in integrin-α M (encoded by ITGAM) is associated with systemic lupus erythematosus.
Nath S K, et al.
Nature Genetics, 40(2), 152-154 (2008)
The pathogenic relevance of ?M-integrin in Guillain?Barre syndrome.
Dong C, et al.
Acta Neuropathologica, 132(5), 739-752 (2016)
Prognostic value of CD11b expression level for acute myeloid leukemia patients: a meta-analysis.
Xu S, et al.
PLoS ONE, 10(8), e0135981-e0135981 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service