Key Documents
ADAPA438107-EP
Adapter for Eppendorf® A-4-38 Rotor
holds 10 x 5-7 mL round-bottom tubes, for use with 90 mL rectangular bucket, pkg of 2 ea
About This Item
UNSPSC Code:
41103911
Pricing and availability is not currently available.
feature
holds 10 x 5-7 mL round-bottom tubes
packaging
pack of 2 ea
pkg of 2 ea
manufacturer/tradename
Eppendorf® 022639323
technique(s)
centrifugation: suitable
compatibility
for use with 90 mL rectangular bucket
General description
Adapter to be used with 90 mL rectangular bucket in Eppendorf A-4-38 rotor. Holds 10 x 5-7 mL round-bottom tubes and supplied as a pack of 2.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)