Skip to Content
Merck
All Photos(2)

Key Documents

A9655

Sigma-Aldrich

Auramine O

Dye content, ≥80%, certified by the Biological Stain Commission, powder

Synonym(s):

4,4′-(Imidocarbonyl)bis(N,N-dimethylaniline) monohydrochloride, Basic Yellow 2, Pyoctaninum aureum

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21N3 · HCl
CAS Number:
Molecular Weight:
303.83
Colour Index Number:
41000
Beilstein:
4030061
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

Product Name

Auramine O, certified by the BSC, Dye content ≥80 %, certified by the Biological Stain Commission

grade

certified by the Biological Stain Commission

Agency

certified by the BSC

form

powder

composition

Dye content, ≥80%

technique(s)

microbe id | staining: suitable

color

yellow to green

mp

>250 °C (dec.) (lit.)

solubility

ethanol: 1 mg/mL, yellow to orange

λmax

370 nm in H2O
432 nm in H2O

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl[H].CN(C)c1ccc(cc1)C(=N)c2ccc(cc2)N(C)C

InChI

1S/C17H21N3.ClH/c1-19(2)15-9-5-13(6-10-15)17(18)14-7-11-16(12-8-14)20(3)4;/h5-12,18H,1-4H3;1H

InChI key

KSCQDDRPFHTIRL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified for use by fluorescence microscopy in Churukian′s modification of Truant′s fluorescent method for acid fast bacilli on paraffin sections.

Auramine O is used for the staining of acid-fast organisms like coccidia.[1] Along with carbol, it generates brilliant yellow fluorochrome of tubercle bacilli. It is effective in detecting positive cases of tuberculosis.[2] Auramine O binds to the mycolic acid in the bacterial cell wall.[3]

Application

Auramine O has been used to study its absorption by a natural biopolymer poly (γ-glutamic acid).[4]

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

E W Koneman.
Koneman's Color Atlas and Textbook of Diagnostic Microbiology, 1565-1565 (2006)
E Kohen.
Cell Structure and Function by Microspectrofluorometry, 490-490 (2014)
Equilibrium and kinetic studies on sorption of basic dyes by a natural biopolymer poly (γ-glutamic acid).
Inbaraj B S, et al.
Biochemical Engineering Journal, 31(3), 204-215 (2006)
Ismael A Heisler et al.
The journal of physical chemistry. B, 113(6), 1623-1631 (2009-01-17)
The effects of confinement on the ultrafast torsional reaction of auramine O in aqueous solution are investigated through ultrafast fluorescence up-conversion with 50 fs time resolution. The aqueous solution is confined in nanoscale water droplets by an ionic surfactant. The
Jessica Minion et al.
Journal of clinical microbiology, 49(5), 2024-2026 (2011-03-25)
Light-emitting diode fluorescence microscopy is being scaled up for tuberculosis control, but fading of auramine-stained slides could compromise external quality assurance. We stored auramine-stained slides and reexamined them over time. Slides stored in all environments faded quickly, with significant changes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service