Skip to Content
Merck

Skip To

A9062

Adenosine 5′-triphosphate di(tris) salt hydrate

≥95%, Fermentation, powder

Synonym(s):

ATP, 5′-ATP-DiTris

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Check Cart for Availability
$56.20
1 g
Check Cart for Availability
$122.00
5 x 1 g
Check Cart for Availability
$490.00
10 g
Check Cart for Availability
$830.00

About This Item

Empirical Formula (Hill Notation):
C10H16N5O13P3 · 2C4H11NO3 · xH2O
CAS Number:
Molecular Weight:
749.45 (anhydrous basis)
MDL number:
UNSPSC Code:
41106305
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.51
Assay:
≥95%
Biological source:
(Fermentation)
Form:
powder
Solubility:
H2O: 50 mg/mL
Storage temp.:
−20°C

$56.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Adenosine 5′-triphosphate di(tris) salt hydrate, ≥95%

biological source

(Fermentation)

Quality Segment

assay

≥95%

form

powder

solubility

H2O: 50 mg/mL

cation traces

Na: ≤0.1%

shipped in

dry ice

storage temp.

−20°C

SMILES string

O.NC(CO)(CO)CO.NC(CO)(CO)CO.Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP(O)(=O)OP(O)(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O

InChI

1S/C10H16N5O13P3.2C4H11NO3.H2O/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(26-10)1-25-30(21,22)28-31(23,24)27-29(18,19)20;2*5-4(1-6,2-7)3-8;/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H,23,24)(H2,11,12,13)(H2,18,19,20);2*6-8H,1-3,5H2;1H2/t4-,6-,7-,10-;;;/m1.../s1

InChI key

ZMNHJZFDZFGOFE-MSQVLRTGSA-N

Application

Adenosine 5′-triphosphate di(tris) salt hydrate has been used:
  • to treat cells for Ca2+ influx measurements in stromal interaction molecule 1 knockout (STIM1−/−) mice and in vitro chemotaxis assay for microglial migration
  • supplemented in buffer C for mitochondrial respiratory control assay and complex V activity assay
  • in the inhibition of silver nano particles (AgNPs) on Na+/K+-ATPase activity

Biochem/physiol Actions

Adenosine 5′-triphosphate di(tris) salt dehydrate is useful in applications that require the absence of metals.
Adenosine 5′-triphosphate (ATP) is a central component of energy storage and metabolism in vivo. ATP is use in many cellular processes, respiration, biosynthetic reactions, motility, and cell division. ATP is a substrate of many kinases involved in cell signaling and of adenylate cyclase(s) that produce the second messenger cAMP. ATP provides the metabolic energy to drive metabolic pumps. ATP serves as a coenzyme in a wide array of enzymatic reactions.
P2 purinergic agonist; increases activity of Ca2+-activated K+ channels; substrate for ATP-dependent enzyme systems

Disclaimer

Unstable: For maximum stability, store as frozen solution at pH 3.5 to 6.8.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A2383A3377A2647
biological source

(Fermentation)

biological source

microbial

biological source

microbial

biological source

natural (organic)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder

form

crystalline

form

powder

assay

≥95%

assay

≥99%

assay

≥97% (HPLC)

assay

≥93% (HPLC)

solubility

H2O: 50 mg/mL

solubility

H2O: 50 mg/mL

solubility

H2O: 50 mg/mL

solubility

H2O: 50 mg/mL

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters