Skip to Content
Merck

Skip To

A8673

Apomyoglobin from equine skeletal muscle

Protein sequencing standard, lyophilized powder

Synonym(s):

ApoMb

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
NACRES:
NA.26
UNSPSC Code:
12352202
MDL number:
Form:
lyophilized powder
Mol wt:
16,951 Da by calculation
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

form

lyophilized powder

mol wt

16,951 Da by calculation

packaging

vial of ~60 nmol

storage temp.

−20°C

Application

  • Identification of carbonylation sites in apomyoglobin after exposure to 4-hydroxy-2-nonenal by solid-phase enrichment and liquid chromatography-electrospray ionization tandem mass spectrometry.: This research identifies specific carbonylation sites in apomyoglobin following exposure to 4-hydroxy-2-nonenal, using advanced mass spectrometry techniques for precise localization of oxidative modifications. (Rauniyar et al., 2010).

  • Analysis of heterogeneous fluorescence decays in proteins. Using fluorescence lifetime of 8-anilino-1-naphthalenesulfonate to probe apomyoglobin unfolding at equilibrium.: This study uses fluorescence lifetime measurements to analyze the unfolding processes of apomyoglobin, providing insights into protein stability and folding dynamics. (Wang et al., 2006).

Preparation Note

Prepared by the method of Rothgeb and Gurd

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A7642A9650P6635
mol wt

16,951 Da by calculation

mol wt

44287 Da by calculation, ~45 kDa

mol wt

1296 Da by calculation (Average Mass)

mol wt

97,200 Da by calculation

form

lyophilized powder

form

powder

form

powder

form

lyophilized powder

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

300

packaging

vial of ~60 nmol

packaging

vial of 25 mg

packaging

-

packaging

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Physical methods for the study of myoglobin.
T M Rothgeb et al.
Methods in enzymology, 52, 473-486 (1978-01-01)
Simonetta Caira et al.
Food chemistry, 230, 482-490 (2017-04-15)
The European reference method (ERM) recognises the fraudulent addition of bovine (B) milk in water buffalo (WB) milk/dairy products based on concomitant isoelectric focusing (IEF) detection of B γ
Thomas Kupke et al.
Communications biology, 3(1), 73-73 (2020-02-16)
Transmembrane signaling proteins play a crucial role in the transduction of information across cell membranes. One function of regulated intramembrane proteolysis (RIP) is the release of signaling factors from transmembrane proteins. To study the role of transmembrane domains (TMDs) in modulating



Global Trade Item Number

SKUGTIN
A8673-1VL04061833400494
A8673-5X1VL04061833400500

Questions

Reviews

No rating value

Active Filters