Skip to Content
Merck
All Photos(1)

Key Documents

A8530

Sigma-Aldrich

m-Aminophenylboronic acid–Agarose

saline suspension

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$242.00

$242.00


Estimated to ship onMay 29, 2025



Select a Size

Change View
5 ML
$242.00

About This Item

MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.32

$242.00


Estimated to ship onMay 29, 2025


form

saline suspension

extent of labeling

5-20 μmol per mL

matrix

cross-linked 6% beaded agarose

matrix activation

epoxy

matrix attachment

amino

matrix spacer

12 atoms

storage temp.

2-8°C

SMILES string

[X]Nc1cc(ccc1)B(O)O

Looking for similar products? Visit Product Comparison Guide

General description

m-Aminophenylboronic acid–Agarose is applicable as a boronate affinity matrix for the purification of severe acute respiratory syndrome′s (SARS-CoV) nucleoprotein (NP) antigen.[1]

Application

m-Aminophenylboronic acid–Agarose has been used as a separating gel in the boronate affinity chromatography of albumins.[2][3] It has also been used as a resin for purification of 3,4-dihydroxyphenylacetaldehyde (DOPAL)-bound α-synuclein (α-syn).[4]

Physical form

Suspension in 0.5 M NaCl, 0.1 M sodium acetate, pH 5.0.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serge Chesne et al.
Biochimie, 88(10), 1467-1477 (2006-07-04)
Non-enzymatic glycosylation (glycation) and oxidative damages represent major research areas insofar as such modifications of proteins are frequently observed in numerous states of disease. Albumin undergoes structural and functional alterations, caused by increased glycosylation during non insulin-dependent diabetes mellitus, which
Philippe Rondeau et al.
Archives of biochemistry and biophysics, 460(1), 141-150 (2007-02-27)
Structural modifications of bovine serum albumin (BSA) induced by heating, and the involvement of glycation of albumin in such processing were studied by using Fourier transform infrared spectroscopy (FTIR) and polyacrylamide gel electrophoresis (PAGE). For native BSA, heating treatments gave
N Plotegher et al.
Scientific reports, 7, 40699-40699 (2017-01-14)
Parkinson's disease is a neurodegenerative disorder characterized by the death of dopaminergic neurons and by accumulation of alpha-synuclein (aS) aggregates in the surviving neurons. The dopamine catabolite 3,4-dihydroxyphenylacetaldehyde (DOPAL) is a highly reactive and toxic molecule that leads to aS
Pravin K Bhatnagar et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 863(2), 235-241 (2008-02-09)
Hybrid hybridomas (quadromas) are derived by fusing at least two hybridomas, each producing a different antibody of predefined specificity. The resulting cell secretes not only the immunoglobulins of both parents but also hybrid molecules manifesting the binding characteristics of the
Francesco Longo et al.
Acta neuropathologica communications, 5(1), 22-22 (2017-03-16)
Mutations in the leucine-rich repeat kinase 2 (LRRK2) gene are the most common genetic cause of Parkinson's disease. Here, we investigated whether the G2019S LRRK2 mutation causes morphological and/or functional changes at nigro-striatal dopamine neurons. Density of striatal dopaminergic terminals

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service