Skip to Content
Merck
All Photos(1)

Key Documents

A7401

Sigma-Aldrich

[Sar1, Gly8]-Angiotensin II acetate salt hydrate

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H65N13O10 · xC2H4O2 · yH2O
Molecular Weight:
912.05 (anhydrous free base basis)
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

storage temp.

−20°C

Amino Acid Sequence

Sar-Arg-Val-Tyr-Ile-His-Pro-Gly

Biochem/physiol Actions

Angiotensin II antagonist

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of peptides with the solid phase method. II. Octapeptide analogues of angiotensin II.
W K Park et al.
Canadian journal of biochemistry, 52(2), 113-119 (1974-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service