Skip to Content
Merck

A7361

Sigma-Aldrich

2R, 4R-APDC monohydrate

≥98% (HPLC)

Synonym(s):

4-amino-2,4-pyrrolidinedicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10N2O4 · H2O
CAS Number:
Molecular Weight:
192.17
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white

solubility

H2O: ≥2 mg/mL

originator

Eli Lilly

storage temp.

room temp

SMILES string

O.N[C@]1(CN[C@H](C1)C(O)=O)C(O)=O

InChI

1S/C6H10N2O4.H2O/c7-6(5(11)12)1-3(4(9)10)8-2-6;/h3,8H,1-2,7H2,(H,9,10)(H,11,12);1H2/t3-,6-;/m1./s1

InChI key

XAIFFDPAANKDRF-VIKFZCHMSA-N

Application

2R,4R-APDC is a Group II mGluR metabotropic glutamate receptor agonist.

Biochem/physiol Actions

2R,4R-APDC is a Group II mGluR metabotropic glutamate receptor.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Glutamate Receptors (G Protein Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Eli Lilly. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service