Skip to Content
Merck
All Photos(1)

Documents

A7255

Sigma-Aldrich

Amyloglucosidase from Rhizopus sp.

greener alternative

≥5,000 units/g solid

Synonym(s):

1,4-α-D-Glucan glucohydrolase, Exo-1,4-α-glucosidase, Glucoamylase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

specific activity

≥5,000 units/g solid

contains

diatomaceous earth, starch and sugar as balance

composition

Protein, ~35% biuret

greener alternative product characteristics

Waste Prevention: Greener alternative product characteristics
Learn more about the Principles of Green Chemistry.

greener alternative category

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for waste prevention when used in starch ethanol research. For more information see the article in biofiles.

Unit Definition

One unit will liberate 1.0 mg of glucose from starch in 3 min at pH 4.5 at 55 °C.

Analysis Note

Using soluble starch, almost theoretical yields of glucose are obtained.

inhibitor

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A F Brito et al.
Journal of dairy science, 91(10), 3968-3982 (2008-10-04)
Alfalfa (Medicago sativa L.) cut at sundown has been shown to contain greater concentration of total nonstructural carbohydrates (TNC) than that cut at sunup. Fourteen multiparous (8 ruminally cannulated) and 2 primiparous lactating dairy cows were randomly assigned to 2
Z Nie et al.
Journal of dairy science, 92(4), 1702-1711 (2009-03-25)
Carbohydrates in forage crops can be divided into neutral detergent-insoluble fiber and neutral detergent-soluble carbohydrates (NDSC); the latter includes organic acids (OA), total ethanol:water-soluble carbohydrates (TESC), starch, and neutral detergent-soluble fiber (NDSF). The accurate and efficient estimation of NDSC in
Ian J Tetlow et al.
Plant physiology, 146(4), 1878-1891 (2008-02-12)
Protein-protein interactions among enzymes of amylopectin biosynthesis were investigated in developing wheat (Triticum aestivum) endosperm. Physical interactions between starch branching enzymes (SBEs) and starch synthases (SSs) were identified from endosperm amyloplasts during the active phase of starch deposition in the
Fushan Liu et al.
Journal of experimental botany, 60(15), 4423-4440 (2009-10-07)
The amylose extender (ae(-)) mutant of maize lacks starch branching enzyme IIb (SBEIIb) activity, resulting in amylopectin with reduced branch point frequency, and longer glucan chains. Recent studies indicate isozymes of soluble starch synthases form high molecular weight complexes with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service