Skip to Content
Merck

Skip To

A6784

Albumin solution human

10% in 0.85% sodium chloride and 0.05% sodium azide, aseptically filled

Synonym(s):

HSA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
50 mL
Check Cart for Availability
$445.00

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.25
MDL number:
eCl@ss:
42010201
Biological source:
human
Form:
liquid
Technique(s):
ELISA: suitable, tissue culture: suitable, western blot: suitable
Impurities:
HIV I and HIVII, HCV and HBsAg, tested negative
Concentration:
10% in 0.85% sodium chloride and 0.05% sodium azide, 9.5-11.4% protein (biuret)

$445.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

sterility

aseptically filled

form

liquid

concentration

10% in 0.85% sodium chloride and 0.05% sodium azide, 9.5-11.4% protein (biuret)

technique(s)

ELISA: suitable, tissue culture: suitable, western blot: suitable

impurities

HIV I and HIVII, HCV and HBsAg, tested negative

UniProt accession no.

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C3F8/c4-1(5,2(6,7)8)3(9,10)11

InChI key

QYSGYZVSCZSLHT-UHFFFAOYSA-N

Gene Information

human ... ALB(213)

General description

Human serum albumin (HSA) is a monomeric, globular, and α-helical protein that constitutes a major part of human blood plasma proteins. This single-chain polypeptide protein contains 585 amino acid residues and 17 internal disulfide bridges and one free cysteine.
Human serum albumin undergoes three different post-translational modifications: oxidation, glycation, and S-nitrosylation. Modifications usually occur on the surface of the globular protein, and do not significantly affect conformation. However, modification strongly affects binding of fatty acids and drug molecules.

Application

Albumin solution human has been used as a supplement in RPMI 1640 media during polymorphonuclear cell (PMN) incubation. It has also been used as a culture media for the differentiation and maturation of monocyte-derived dendritic cells.

Biochem/physiol Actions

Human serum albumin (HSA) plays a vital role in the regulation of the colloidal osmotic pressure of blood. It also aids in the maintenance of metal ion homeostasis, including the transport and storage of transition metals. The high binding capacity of HSA with a wide range of ions and molecules makes it an effective molecular cargo and nano vehicle used in biophysical, clinical, and industrial fields. Human and bovine albumins contain 16% nitrogen and are often used as standards in protein calibration studies. It is used as a blocking agent in Western blots or enzyme-linked immunosorbent assay (ELISA) applications. Globulin-free albumins are suitable for use in applications where no other proteins should be present (e.g., electrophoresis).

Features and Benefits

  • Easily crystallized and contain an excess of acidic amino acids.
  • Serum and plasma albumin is carbohydrate-free and comprises 55-62% of the protein present
  • Due to its high charge to mass ratio albumin binds water, Ca2+, Na+, K+, fatty acids, bilirubin, hormones, and drugs.
  • The free hydrophobic region of fatty acid-free albumins helps to solubilize lipids in tissue culture

Other Notes

View more information on human serum albumin.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A3782A9511A6608
technique(s)

ELISA: suitable, western blot: suitable, tissue culture: suitable

technique(s)

ELISA: suitable, tissue culture: suitable, western blot: suitable

technique(s)

ELISA: suitable, tissue culture: suitable, western blot: suitable

technique(s)

cell culture | mammalian: suitable

biological source

human

biological source

human

biological source

human

biological source

human

Quality Level

200

Quality Level

200

Quality Level

300

Quality Level

200

form

liquid

form

lyophilized powder

form

lyophilized powder

form

aqueous solution

sterility

aseptically filled

sterility

-

sterility

-

sterility

-

Gene Information

human ... ALB(213)

Gene Information

human ... ALB(213)

Gene Information

human ... ALB(213)

Gene Information

human ... ALB(213)


Still not finding the right product?

Explore all of our products under Albumin solution human


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
A6784-10ML04061837145988
A6784-5ML04061837145995
A6784-50ML04061833382844

Questions

  1. Добры й день !Подскажите пожалуйста срок годности albumin solution human-10% in 0.85%sodium chloride A6784, SCLF8390

    1 answer
    1. his product is not assigned an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one year warranty from the date of shipment. For more information you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

Reviews

No rating value

Active Filters