Skip to Content
Merck

A6720

N-Acetyl-Trp-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin trifluoroacetate salt

≥97% (HPLC), powder

Synonym(s):

Ac-WEHD-AFC

Sign In to View Organizational & Contract Pricing.

Select a Size

5 MG

$677.00

$677.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

About This Item

NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352209
MDL number:
Assay:
≥97% (HPLC)
Form:
powder
Quality level:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

assay

≥97% (HPLC)

form

powder

color

yellow

solubility

DMSO/DMF: 20 mM

storage temp.

−20°C

SMILES string

CC(=O)N[C@@H](Cc1c[nH]c2ccccc12)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](Cc3c[nH]cn3)C(=O)N[C@@H](CC(O)=O)C(=O)Nc4ccc5c(OC(=O)C=C5C(F)(F)F)c4

InChI

1S/C38H37F3N8O11/c1-18(50)45-27(10-19-15-43-25-5-3-2-4-22(19)25)36(58)47-26(8-9-31(51)52)34(56)48-28(11-21-16-42-17-44-21)37(59)49-29(14-32(53)54)35(57)46-20-6-7-23-24(38(39,40)41)13-33(55)60-30(23)12-20/h2-7,12-13,15-17,26-29,43H,8-11,14H2,1H3,(H,42,44)(H,45,50)(H,46,57)(H,47,58)(H,48,56)(H,49,59)(H,51,52)(H,53,54)/t26-,27-,28-,29-/m0/s1

InChI key

NEGOLAGPKZGIKJ-DZUOILHNSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
A1086A0466A9965
assay

≥97% (HPLC)

assay

≥97% (HPLC)

assay

≥90% (HPLC)

assay

≥95% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

200

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

solubility

DMSO/DMF: 20 mM

solubility

DMSO: 10 mM

solubility

H2O: 1 mg/mL

solubility

-

color

yellow

color

white

color

white

color

white

Biochem/physiol Actions

Fluorogenic substrate for caspase 1 and 5.

Features and Benefits

This compound is featured on the Caspases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T A Rano et al.
Chemistry & biology, 4(2), 149-155 (1997-02-01)
Interleukin-1beta converting enzyme (ICE/caspase-1) is the protease responsible for interleukin-1beta (IL-1beta) production in monocytes. It was the first member of a new cysteine protease family to be identified. Members of this family have functions in both inflammation and apoptosis. A
R V Talanian et al.
The Journal of biological chemistry, 272(15), 9677-9682 (1997-04-11)
The caspase family represents a new class of intracellular cysteine proteases with known or suspected roles in cytokine maturation and apoptosis. These enzymes display a preference for Asp in the P1 position of substrates. To clarify differences in the biological

Related Content

Global Trade Item Number

SKUGTIN
A6720-5MG04061833267547

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service