Skip to Content
Merck
All Photos(1)

Key Documents

A6486

Sigma-Aldrich

AS-136A

≥98% (HPLC)

Synonym(s):

1-Methyl-N-[4-(1-piperidinylsulfonyl)phenyl]-3-(trifluoromethyl)-1H-pyrazole-5-carboxamide, CID 16122506, SID 24769845;

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19F3N4O3S
CAS Number:
Molecular Weight:
416.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

Cn1nc(cc1C(=O)Nc2ccc(cc2)S(=O)(=O)N3CCCCC3)C(F)(F)F

InChI

1S/C17H19F3N4O3S/c1-23-14(11-15(22-23)17(18,19)20)16(25)21-12-5-7-13(8-6-12)28(26,27)24-9-3-2-4-10-24/h5-8,11H,2-4,9-10H2,1H3,(H,21,25)

InChI key

OJDLQNVQEAZKSL-UHFFFAOYSA-N

Application

AS-136A is a small molecule developed as part of the NIH Probes effort. AS-136A is a potent measles virus (MeV) RNA-dependent RNA polymerase (RdRp) inhibitor. AS-136A is very stable, decreases virus titer after 24 hours in physiological conditions, and serves as a positive control in MeV inhibitory assays.

Biochem/physiol Actions

AS-136A is a potent measles virus RdRp inhibitor. Measles virus (MV) is a member of the paramyxovirus family. Ribavirin (R9644) is the only drug available for treatment of paramyxoviruses, and it has limited efficacy against MV. A series of small molecules has been developed to inhibit viral RNA-dependent RNA polymerase (RdRp), a protein vital to infection leading to the development of AS-136A, a very potent RdRp inhibitor. AS-136A has high stability and continues to decrease virus titer after 24 hours in physiologic conditions. AS-136A is now used as a positive control in experiments assaying MV inhibition. This compound is a product of the NIH Probes effort, also known as SID 24769845.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeong-Joong Yoon et al.
Antimicrobial agents and chemotherapy, 53(9), 3860-3870 (2009-06-17)
No effective therapeutic is currently in place for improved case management of severe measles or the rapid control of outbreaks. Through high-throughput screening, we recently identified a novel small-molecule class that potently blocks activity of the measles virus (MeV) RNA-dependent
J Maina Ndungu et al.
Journal of medicinal chemistry, 55(9), 4220-4230 (2012-04-07)
The measles virus (MeV), a member of the paramyxovirus family, is an important cause of pediatric morbidity and mortality worldwide. In an effort to provide therapeutic treatments for improved measles management, we previously identified a small, non-nucleoside organic inhibitor of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service