Skip to Content
Merck
All Photos(1)

Key Documents

A5972

Sigma-Aldrich

Acetyl-Diabetes Associated Peptide Fragment 8-37 amide trifluoroacetate salt

sequence human, ≥95% (HPLC)

Synonym(s):

Acetyl-Amylin Fragment 8-37 amide trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 MG
$453.00

$453.00


Estimated to ship onApril 17, 2025



Select a Size

Change View
0.5 MG
$453.00

About This Item

Empirical Formula (Hill Notation):
C140H218N42O46 · xC2HF3O2
Molecular Weight:
3225.48 (free base basis)
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.32

$453.00


Estimated to ship onApril 17, 2025


biological source

sequence from human

Quality Level

Assay

≥95% (HPLC)

form

solid

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IAPP(3375)

Amino Acid Sequence

Ac-Ala-Thr-Gln-Arg-Leu-Ala-Asn-Phe-Leu-Val-His-Ser-Ser-Asn-Asn-Phe-Gly-Ala-Ile-Leu-Ser-Ser-Thr-Asn-Val-Gly-Ser-Asn-Thr-Tyr-NH2

Biochem/physiol Actions

Reverses the inhibitory effects of amylin on glycogen accumulation in vitro

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service