Skip to Content
Merck
All Photos(1)

Key Documents

A5756

Sigma-Aldrich

L-Arginine L-glutamate salt

Synonym(s):

S(+)-2-Amino-5-guanidinopentanoic acid L-glutamate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H23N5O6
CAS Number:
Molecular Weight:
321.33
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

mol wt

321.3

storage temp.

−20°C

SMILES string

N[C@@H](CCC(O)=O)C(O)=O.O=C(O)[C@@H](N)CCCNC(N)=N

InChI

1S/C6H14N4O2.C5H9NO4/c7-4(5(11)12)2-1-3-10-6(8)9;6-3(5(9)10)1-2-4(7)8/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);3H,1-2,6H2,(H,7,8)(H,9,10)/t4-;3-/m00/s1

InChI key

RVEWUBJVAHOGKA-WOYAITHZSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO). Induces insulin release by a nitric oxide-dependent mechanism.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prebiotic evolution: an approach to polymerisation based on crystal structures.
M Vijayan
FEBS letters, 112(2), 135-137 (1980-04-07)
I I Krychun
Likars'ka sprava, (3-4)(3-4), 49-56 (2009-01-17)
It has been established that the blood content of protein P53 diminishes by 27%, the blood level of sTRAIL increases by 22%, sCD 117 increases by 44% in patients with vegeto-vascular dystonia of the hypertonic type that is accompanied by
T Nagamine et al.
Journal of gastroenterology, 30(3), 351-355 (1995-06-01)
The effects of biotin on ammonia concentration in blood and brain were evaluated in hyperammonemic rats and mice. Rats were injected with 5 mmol/kg BW of ammonium acetate, and mice were injected with 10 mmol/kg BW. Increases in blood ammonia
[Chronic liver disease].
Akinori Ueki
Ryoikibetsu shokogun shirizu, (40)(40), 197-200 (2003-11-25)
Thierry Lebret et al.
European urology, 41(6), 608-613 (2002-06-21)
The goal of this double-blind, placebo-controlled, three-way crossover, randomized clinical trial was to compare the efficacy and safety of the combination of 6g of L-arginine glutamate and 6 mg of yohimbine hydrochloride (AY) with that of 6 mg of yohimbine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service