Skip to Content
Merck
All Photos(1)

Key Documents

A5330

Sigma-Aldrich

N-Acetyl-L-tryptophan 3,5-bis(trifluoromethyl)benzyl ester

≥99% (TLC), powder

Synonym(s):

L-732,138

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H18F6N2O3
CAS Number:
Molecular Weight:
472.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99% (TLC)

form

powder

mp

147-148 °C (lit.)

storage temp.

−20°C

SMILES string

CC(=O)N[C@@H](Cc1c[nH]c2ccccc12)C(=O)OCc3cc(cc(c3)C(F)(F)F)C(F)(F)F

InChI

1S/C22H18F6N2O3/c1-12(31)30-19(8-14-10-29-18-5-3-2-4-17(14)18)20(32)33-11-13-6-15(21(23,24)25)9-16(7-13)22(26,27)28/h2-7,9-10,19,29H,8,11H2,1H3,(H,30,31)/t19-/m0/s1

InChI key

BYYQYXVAWXAYQC-IBGZPJMESA-N

Gene Information

human ... TACR1(6869)

Biochem/physiol Actions

Competitive substance P receptor antagonist.
Competitive substance P receptor antagonist. It is approximately 1000-fold more potent at cloned human NK-1 receptors than at cloned human NK-2 and NK-3 receptors; and approximately 200-fold more potent at human NK-1 receptors than at rat NK-1 receptors. The IC50 for the human NK-1 receptor expressed in CHO cells is approximately 2 nM.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takenori Onaga et al.
Regulatory peptides, 173(1-3), 64-73 (2011-10-06)
The present study investigated a role of tachykinins (TK) and neurokinin (NK) receptors (NK-R) in the non-cholinergic regulation of omasal contractions in sheep. Semiquantitative reverse transcription (RT)-PCR revealed that both preprotachykinin (PPT)-A and PPT-B mRNA were distributed in the omasal
Ammar Boudaka et al.
European journal of pharmacology, 556(1-3), 157-165 (2006-12-13)
Transient receptor potential ion channel of the vanilloid type 1 (TRPV1)-dependent pathway, consisting of capsaicin-sensitive tachykininergic primary afferent and myenteric nitrergic neurons, has been suggested to mediate the inhibitory effect of capsaicin on vagally mediated striated muscle contractions in the
Ming-Ming Zhang et al.
PloS one, 8(3), e59234-e59234 (2013-04-05)
Substance P (SP) and its receptor, the neurokinin 1 receptor (NK1R), play important roles in transmitting and regulating somatosensory nociceptive information. However, their roles in visceral nociceptive transmission and regulation remain to be elucidated. In the previous study, moderate SP
Miguel Muñoz et al.
Investigational new drugs, 26(2), 111-118 (2007-10-02)
It has been demonstrated that substance P (SP) induces cell proliferation and neurokinin-1 (NK-1) receptor antagonists inhibit growth in several human cancer cell lines, but it is currently unknown whether such actions are exerted on human laryngeal carcinoma cell line
Miguel Muñoz et al.
Peptides, 38(2), 318-325 (2012-10-03)
The last decades have seen no significant progress in extending the survival of lung cancer patients and there is an urgent need to improve current therapies. The substance P (SP)/neurokinin-1 receptor (NK-1R) system plays an important role in the development

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service