Skip to Content
Merck
All Photos(1)

Key Documents

A5284

Sigma-Aldrich

8-Azaguanine

Hybri-Max, γ-irradiated, powder, BioXtra, suitable for hybridoma

Synonym(s):

8-AzaG, 2-Amino-6-hydroxy-8-azapurine, 2-Amino-6-oxy-8-azapurine, Guanazolo

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N6O
CAS Number:
Molecular Weight:
152.11
Beilstein:
171098
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:

grade

Hybri-Max

sterility

γ-irradiated

product line

BioXtra

form

powder

technique(s)

cell culture | hybridoma: suitable

mp

>300 °C (lit.)

storage temp.

−20°C

SMILES string

NC1=Nc2[nH]nnc2C(=O)N1

InChI

1S/C4H4N6O/c5-4-6-2-1(3(11)7-4)8-10-9-2/h(H4,5,6,7,8,9,10,11)

InChI key

LPXQRXLUHJKZIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For use in hybridoma cell culture applications as a myeloma selection agent.

Reconstitution

Reconstitute contents of vial with 10 mL sterile cell culture medium. Stock solution is sufficient to prepare 500 mL medium.

Legal Information

Hybri-Max is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

July Fong et al.
Scientific reports, 8(1), 1155-1155 (2018-01-20)
The threat of antibiotic resistant bacteria has called for alternative antimicrobial strategies that would mitigate the increase of classical resistance mechanism. Many bacteria employ quorum sensing (QS) to govern the production of virulence factors and formation of drug-resistant biofilms. Targeting
F R Ridzlan et al.
Tropical biomedicine, 27(3), 632-638 (2011-03-15)
Leptospirosis is recognized as one of the important zoonotic diseases in the world including Malaysia. A total of 145 soil and water samples were collected from selected National Service Training Centres (NSTC) in Kelantan and Terengganu. The samples were inoculated
S Grenacher et al.
Experimental & applied acarology, 25(8), 641-660 (2002-08-13)
Ticks are ectoparasites of vertebrates and utilize a variety of infochemicals for host finding and acceptance as well as for intraspecific aggregation and mating responses. Individual male and female Ixodes ricinus. the vector of Lyme disease in Europe. readily arrest
Bashir Mir et al.
Nucleic acids research, 32(3), e25-e25 (2004-02-13)
The use of primary somatic cells in nuclear transfer procedure has opened a new opportunity to manipulate domestic animal genomes via homologous recombination. To date, while a few loci have been targeted in somatic cells using similar enrichment strategies as
Ziwei Li et al.
International journal of oncology, 52(6), 2079-2092 (2018-04-06)
Hepatocellular carcinoma (HCC) is one of the most common malignancies and causes of death worldwide. Research investigating novel therapeutic strategies for the treatment of HCC is urgently required. Monoclonal antibodies (mAbs) that target the programmed cell death‑1 (PD‑1/PDCD1)/programmed death-ligand 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service