Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
100 mg | Check Cart for Availability | $121.00 |
500 mg | Check Cart for Availability | $353.00 |
1 g | Check Cart for Availability | $637.00 |
About This Item
CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-564-0
MDL number:
$121.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Application
L-amino acid oxidase is used to convert L-amino acids to their corresponding α-keto acids. Product A5147 is from Crotalus atrox. L-amino acid oxidase, from Sigma, has been used in leucine aminopeptidase (LAP) activity assays [1].
Biochem/physiol Actions
L-amino acid oxidase is involved in various metabolic pathways such as alanine and aspartate metabolism, methionine metabolism, valine, leucine and isoleucine degradation, tyrosine metabolism, phenylalanine metabolism, tryptophan metabolism, phenylalanine, tyrosine and tryptophan biosynthesis, and alkaloid biosynthesis [2].
1 of 1
This Item | |||
|---|---|---|---|
| form dried venom | form crude dried venom | form powder | form aqueous suspension |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. 2-8°C |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| type Type VI | type Type IV | type Type I (dried venom) | type Type IV |
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




