Skip to Content
Merck
All Photos(1)

Documents

A5111

Sigma-Aldrich

ABT-724 trihydrochloride

≥98% (HPLC)

Synonym(s):

2-[[4-(2-Pyridinyl)-1-piperazinyl]methyl]-1H-benzimidazole trihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19N5 · 3HCl
CAS Number:
Molecular Weight:
402.75
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white

solubility

H2O: >30 mg/mL

originator

Abbott

storage temp.

2-8°C

SMILES string

Cl.Cl.Cl.C1CN(CCN1Cc2nc3ccccc3[nH]2)c4ccccn4

InChI

1S/C17H19N5.3ClH/c1-2-6-15-14(5-1)19-16(20-15)13-21-9-11-22(12-10-21)17-7-3-4-8-18-17;;;/h1-8H,9-13H2,(H,19,20);3*1H

InChI key

AZFUVPBLKQGSRI-UHFFFAOYSA-N

Application

ABT-724 is a dopamine D4 receptor agonist. ABT-724 has been used in the study of impulsive behavior and penile erection in rats.

Features and Benefits

This compound is featured on the Dopamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mikhail N Koffarnus et al.
Behavioural pharmacology, 22(4), 300-311 (2011-06-23)
Impulsivity is widely regarded as a multidimensional trait that encompasses two or more distinct patterns of behavior, and dopaminergic systems are implicated in the expression of impulsive behavior in both humans and animal subjects. Impulsive choice, or the tendency to
Ronan Depoortère et al.
Behavioural pharmacology, 20(4), 303-311 (2009-07-09)
Dopamine (DA) is implicated in penile erection (PE) and yawning (YA) in rats through activation of D2-like receptors. However, the exact role of each subtype (D2, D3 and D4) of this receptor family in PE/YA is still not clearly elucidated.
Danuta Marona-Lewicka et al.
Psychopharmacology, 203(2), 265-277 (2008-07-08)
Lysergic acid diethylamide (LSD) differs from other types of hallucinogens in that it possesses direct dopaminergic effects. The exact nature of this component has not been elucidated. The present study sought to characterize the effects of several dopamine D(4) agonists

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service