A4963
Annexin V Cy3.18 Conjugate from human placenta
buffered aqueous solution
Synonym(s):
Annexin V, Calphosbindin I, Lipocortin V, PAP-1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106305
Recommended Products
form
buffered aqueous solution
Quality Level
extent of labeling
1-3 mol Cy3.18 per mol annexin V
UniProt accession no.
shipped in
wet ice
storage temp.
2-8°C
Gene Information
human ... ANXA5(308)
Looking for similar products? Visit Product Comparison Guide
Application
For detection of apoptotic cells by fluorescence microscopy.
Biochem/physiol Actions
Annexins are ubiquitous homologous proteins that bind phospholipids in the presence of calcium.
Annexins are ubiquitous homologous proteins that bind phospholipids in the presence of calcium. The cellular changes involved in the apoptotic process include loss of phospholipid asymmetry during the early stages. This phenomenon is universal and is not limited to stimulus or to mammalian cells, but also occurs in insect and plant cells. In living cells, phosphatidylserine is transported to the inside of the lipid bilayer by the Mg2+ATP dependent enzyme, aminophospholipid translocase. At the onset of apoptosis, phosphatidylserine becomes translocated to the external surface of the cell membrane. Since the movement of phosphatidylserine from the internal membrane surface to the external surface is an early indicator of apoptosis, annexin V and its conjugates that interact strongly and specifically with phosphatidylserine may be used to detect apoptosis. Annexin V conjugates can be used to detect apoptotic cells significantly earlier than DNA-based assays. Fluorescent dye labeled annexin V have applications in flow cytometry, fluorescence microscopy, and laser scanning cytometry.
Physical form
Solution in 50 mM Tris-HCl, pH 7.5, containing 100 mM NaCl.
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Benedikt Linder et al.
International journal of molecular sciences, 23(16) (2022-08-27)
While the fungal metabolite illudin M (1) is indiscriminately cytotoxic in cancer and non-malignant cells, its retinoate 2 showed a greater selectivity for the former, especially in a cerebral context. Illudin M killed malignant glioma cells as well as primary
Adam M Nagy et al.
Biochimica et biophysica acta. Bioenergetics, 1859(3), 201-214 (2017-12-24)
Microglia are highly dynamic cells in the brain. Their functional diversity and phenotypic versatility brought microglial energy metabolism into the focus of research. Although it is known that microenvironmental cues shape microglial phenotype, their bioenergetic response to local nutrient availability
Katryna Pampuscenko et al.
Journal of neurochemistry, 154(3), 316-329 (2019-12-14)
Tau is a microtubule-associated protein, found at high levels in neurons, and its aggregation is associated with neurodegeneration. Recently, it was found that tau can be actively secreted from neurons, but the effects of extracellular tau on neuronal viability are unclear.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service