Skip to Content
Merck

Skip To

A4802

Methyl 1-aminocyclopropanecarboxylate hydrochloride

≥98% (TLC)

Synonym(s):

1-Aminocyclopropanecarboxylic acid methyl ester hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Check Cart for Availability
$363.00

About This Item

Empirical Formula (Hill Notation):
C5H9NO2 · HCl
CAS Number:
Molecular Weight:
151.59
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26
MDL number:

$363.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Methyl 1-aminocyclopropanecarboxylate hydrochloride,

assay

≥98% (TLC)

form

powder

color

white

storage temp.

2-8°C

SMILES string

Cl.COC(=O)C1(N)CC1

InChI

1S/C5H9NO2.ClH/c1-8-4(7)5(6)2-3-5;/h2-3,6H2,1H3;1H

InChI key

CHSDCWKLSHPKFY-UHFFFAOYSA-N

Biochem/physiol Actions

Methyl 1-aminocyclopropanecarboxylate (Methyl-ACC) is a methylated 1-aminocyclopropanecarboxylate which is produced in plants by the enzyme 1-aminocyclopropane-1-carboxylate synthase (ACC synthase).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A5575728934MNO000004
form

powder

form

powder

form

powder

form

solid

assay

≥98% (TLC)

assay

≥98% (TLC)

assay

97%

assay

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

-

color

white

color

white

color

-

color

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library