A4730
Amthamine dihydrobromide
≥98% (HPLC), solid
Synonym(s):
2-Amino-4-methyl-5-thiazoleethanamine dihydrobromide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
≥98% (HPLC)
form
solid
color
off-white
solubility
H2O: 27 mg/mL
storage temp.
2-8°C
SMILES string
Br[H].Br[H].Cc1nc(N)sc1CCN
InChI
1S/C6H11N3S.2BrH/c1-4-5(2-3-7)10-6(8)9-4;;/h2-3,7H2,1H3,(H2,8,9);2*1H
InChI key
XFXNNOPUDSFVJE-UHFFFAOYSA-N
Application
Amthamine dihydrobromide is a histamine H2 receptor agonist. Amthamine dihydrobromide has been used to study the effect of mast cells on T regulatory cell function.
Biochem/physiol Actions
Amthamine dihydrobromide is a H2 histamine receptor agonist. Amthamine dihydrobromide, similar to histamine, inhibits H2 receptor-mediated eosinophil peroxidase (EPO) release with IC50 = 0.4 μM; a weak antagonist at H3 and shows no activity at H1 receptors.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Naunyn-Schmiedeberg's archives of pharmacology, 348(1), 77-81 (1993-07-01)
The new histamine H2 receptor agonist amthamine, [2-amino-5-(2-aminoethyl)-4-methylthiazole], was tested for its activity on gastric acid secretion in different in vivo and in vitro experimental models. Amthamine induced a dose-related increase in acid secretion both in conscious cats with a
Molecular pharmacology, 44(4), 886-894 (1993-10-01)
Recently we developed amthamine [2-amino-5-(2-aminoethyl)-4-methylthiazole]. This cyclic analogue of dimaprit proved to be the most potent and selective histamine H2 receptor agonist of a series of substituted 4- or 5-(2-aminoethyl)thiazoles. Quantum chemical studies on histamine (N pi-H tautomer), dimaprit, and
Journal of medicinal chemistry, 35(17), 3239-3246 (1992-08-21)
It is well known that both histamine and dimaprit show moderate histamine H2-receptor agonistic activities on the guinea pig right atrium. Quantum chemical calculations on these two compounds showed similarities in electron distributions and molecular electrostatic potentials (MEP's), which could
Agents and actions, 40(1-2), 44-49 (1993-09-01)
The cardiac activity of the novel histamine H2-receptor agonist amthamine was investigated in a variety of isolated heart preparations from guinea pigs and humans and in the isolated rabbit aorta. Amthamine caused an increase in the sinus rate of spontaneously
Cardiac effects of amthamine: a new histamine H2-receptor agonist.
European journal of clinical investigation, 25 Suppl 1, 27-28 (1995-03-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service