Skip to Content
Merck
All Photos(1)

Key Documents

A4480

Sigma-Aldrich

Anti-ASPP2 antibody, Mouse monoclonal

~1.5 mg/mL, clone DX 54.10, purified from hybridoma cell culture

Synonym(s):

Anti-53BP2 apoptosis-stimulating protein of p53 2, Anti-TP53BP2, Anti-tumor protein p53-binding protein 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

DX 54.10, monoclonal

form

buffered aqueous solution

mol wt

antigen ~175 kDa

species reactivity

human, mouse

concentration

~1.5 mg/mL

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
microarray: suitable
western blot: 0.5-1 μg/mL using total cell extract of human osteogenic sarcoma

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Monoclonal Anti-ASPP2 (mouse IgG1 isotype) is derived from the hybridoma DX 54.10 produced by the fusion of mouse myeloma cells (SP2/0 cells) and splenocytes from BALB/c mice. Apoptosis-stimulating protein of p53 (ASPP) family of proteins contains three members, ASPP1, ASPP2 and iASPP, which interact via their C-terminal region with p53 and modulate its activity.

Specificity

Monoclonal Anti-ASPP2 recognizes human and mouse ASPP2. The epitope recognized by the antibody resides within amino acids 691-1128 of human ASPP2.

Immunogen

fragment of human ASPP2 (amino acids 691-1128).

Application

Mouse Monoclonal Anti-ASPP2 has been used for immunofluorescence analysis. The antibody can also be used for immunocytochemistry, immunoprecipitation and microarray applications.
Western blot analysis of Hek293 or 3T3-L1 adipocyte cell lysates was performed using monoclonal mouse anti-ASPP2 as the primary.

Biochem/physiol Actions

ASPP2 is a tumor suppressor that activates p53. ASPP2 also regulates Par-3 which subsequently modulates the proliferation and polarity of neural progenitor cells during CNS development. Furthermore, ASPP2 interacts with active Ras to induce p53 mediated cell death. ASPP2 is known to function as a biomarker for invasive pituitary adenomas.
Apoptosis-stimulating protein of p53 (ASPP2) enhances the ability of p53 to induce apoptosis ASPP1 and ASPP2 stimulates the transactivation function of p63 and p73 on promoters of different genes such as BCL2 associated X, apoptosis regulator (Bax), p53-inducible gene 3 (PIG3) and p53 upregulated modulator of apoptosis (PUMA), but not mouse double minute 2 homolog (Mdm2) or p21WAF-1/CIP1.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberta Sottocornola et al.
Developmental cell, 19(1), 126-137 (2010-07-14)
Cell polarity plays a key role in the development of the central nervous system (CNS). Interestingly, disruption of cell polarity is seen in many cancers. ASPP2 is a haplo-insufficient tumor suppressor and an activator of the p53 family. In this
Lin Ma et al.
Oncology letters, 6(5), 1313-1318 (2013-11-02)
Nucleostemin is a GTP-conjugated protein located in the nucleoli of stem cells and certain cancer cells, and maintains cellular self-renewal. The present study aimed to evaluate nucleostemin as a potential target for pituitary adenoma gene therapy by investigating nucleostemin and
Wei-Tzu Sun et al.
Biochemical and biophysical research communications, 376(2), 395-398 (2008-09-17)
The p53 tumor suppressor functions in maintaining the integrity of the genome. We have previously reported that DDA3 is an oncoprotein transcriptionally regulated by p53. To explore mechanisms underlying DDA3 action, we searched for its interacting proteins by yeast two-hybrid
Y Wang et al.
Cell death and differentiation, 20(4), 525-534 (2013-02-09)
RAS mutations occur frequently in human cancer and activated RAS signalling contributes to tumour development and progression. Apart from its oncogenic effects on cell growth, active RAS has tumour-suppressive functions via its ability to induce cellular senescence and apoptosis. RAS
Libin Yin et al.
Oncology letters, 15(5), 6409-6417 (2018-05-08)
The apoptosis-stimulating protein of p53 (ASPP) family is a newly identified family protein including ASPP1, ASPP2 and inhibitor of ASPP (iASPP), by which the tumor protein 53 (TP53)-mediated apoptotic process is selectively regulated. Downregulation of ASPP1/ASPP2 and upregulation of iASPP

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service